siRNA Details
| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106649.1 | NP_001100119.1 | 11 | 1589 | ||
| XM_006238177.2 | XP_006238239.1 | 11 | 2706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnnal1| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106649.1 | NP_001100119.1 | 11 | 1589 | ||
| XM_006238177.2 | XP_006238239.1 | 11 | 2706 | |||