siRNA Details
| Gene Symbol: Olr853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000398.1 | NP_001000398.1 | 1 | 406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr853| Gene Symbol: Olr853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000398.1 | NP_001000398.1 | 1 | 406 | ||