siRNA Details
| Gene Symbol: Olr841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000405.1 | NP_001000405.1 | 1 | 504 | ||
| Gene Symbol: LOC500460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749941.1 | XP_003749989.1 | 1 | 504 | ||
More Information
Set Membership: |
|

Olr841
Genomic Map