siRNA Details
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201271.1 | NP_958428.1 | 2 | 216 | ||
GenBank mRNA | AY299399.1 | - | 2 | 216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201271.1 | NP_958428.1 | 2 | 216 | ||
GenBank mRNA | AY299399.1 | - | 2 | 216 |