siRNA Details
| Gene Symbol: RGD1305420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025652.1 | NP_001020823.1 | 4 | 535 | ||
| XM_008763696.1 | XP_008761918.1 | 4 | 602 | |||
| GenBank mRNA | BC097981.1 | - | 4 | 535 | 7443366 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305420| Gene Symbol: RGD1305420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025652.1 | NP_001020823.1 | 4 | 535 | ||
| XM_008763696.1 | XP_008761918.1 | 4 | 602 | |||
| GenBank mRNA | BC097981.1 | - | 4 | 535 | 7443366 | |