siRNA Details
| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011975.1 | NP_001011975.1 | 2 | 1583 | ||
| GenBank mRNA | BC081884.1 | - | 2 | 1583 | 7108511 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1b1| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011975.1 | NP_001011975.1 | 2 | 1583 | ||
| GenBank mRNA | BC081884.1 | - | 2 | 1583 | 7108511 | |