siRNA Details
| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033693.2 | NP_001028865.2 | 3 | 334 | ||
| XM_006238230.2 | XP_006238292.1 | 2 | 302 | |||
| GenBank mRNA | BC099751.1 | - | 3 | 307 | 5623519 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc31a2| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033693.2 | NP_001028865.2 | 3 | 334 | ||
| XM_006238230.2 | XP_006238292.1 | 2 | 302 | |||
| GenBank mRNA | BC099751.1 | - | 3 | 307 | 5623519 | |