siRNA Details
Gene Symbol: Prpf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106659.1 | NP_001100129.1 | 3 | 433 | ||
GenBank mRNA | BC087165.1 | - | 3 | 375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prpf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106659.1 | NP_001100129.1 | 3 | 433 | ||
GenBank mRNA | BC087165.1 | - | 3 | 375 |