siRNA Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011976.2 | NP_001011976.2 | 6 | 524 | ||
| XM_006238237.2 | XP_006238299.1 | 6 | 488 | |||
| XM_006238239.2 | XP_006238301.1 | 6 | 550 | |||
| GenBank mRNA | BC079286.1 | - | 6 | 524 | 7134286 | |

Wdr31
Genomic Map