siRNA Details
| Gene Symbol: RGD1563975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195270.1 | NP_001182199.1 | 1 | 1527 | ||
| GenBank mRNA | BC128763.1 | - | 1 | 1527 | 7113360 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563975| Gene Symbol: RGD1563975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195270.1 | NP_001182199.1 | 1 | 1527 | ||
| GenBank mRNA | BC128763.1 | - | 1 | 1527 | 7113360 | |