siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039009.1 | NP_001034098.1 | 5, 6 | 694 | ||
| XM_008763765.1 | XP_008761987.1 | 5, 6 | 3596 | |||
| XM_008763766.1 | XP_008761988.1 | 6, 7 | 892 | |||
| XM_008763767.1 | XP_008761989.1 | 6, 7 | 751 | |||
| XM_008763768.1 | XP_008761990.1 | 6, 7 | 3305 | |||
| GenBank mRNA | AY886536.1 | - | 5, 6 | 694 | ||

Zdhhc21
Genomic Map