siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039009.1 | NP_001034098.1 | 2 | 247 | ||
| XM_008763765.1 | XP_008761987.1 | 2 | 3149 | |||
| XM_008763766.1 | XP_008761988.1 | 3 | 445 | |||
| XM_008763767.1 | XP_008761989.1 | 3 | 304 | |||
| XM_008763768.1 | XP_008761990.1 | 3 | 2858 | |||
| GenBank mRNA | AY886536.1 | - | 2 | 247 | ||

Zdhhc21
Genomic Map