siRNA Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013931.1 | NP_001013953.1 | 3 | 843 | ||
| GenBank mRNA | BC087107.1 | - | 3 | 843 | 7193207 | |
| FQ212427.1 | - | 861 | ||||
| FQ212481.1 | - | 861 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam214b| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013931.1 | NP_001013953.1 | 3 | 843 | ||
| GenBank mRNA | BC087107.1 | - | 3 | 843 | 7193207 | |
| FQ212427.1 | - | 861 | ||||
| FQ212481.1 | - | 861 | ||||