siRNA Details
Gene Symbol: Fam214b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013931.1 | NP_001013953.1 | 3, 4 | 1156 | ||
GenBank mRNA | BC087107.1 | - | 3, 4 | 1156 | 7193207 |

More Information
Other Species Targeted

