siRNA Details
Gene Symbol: Stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031646.1 | NP_001026816.1 | 3 | 268 | ||
GenBank mRNA | BC099164.1 | - | 3 | 268 | 7382023 | |
FQ213146.1 | - | 3 | 299 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031646.1 | NP_001026816.1 | 3 | 268 | ||
GenBank mRNA | BC099164.1 | - | 3 | 268 | 7382023 | |
FQ213146.1 | - | 3 | 299 |