siRNA Details
| Gene Symbol: Mysm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238409.2 | XP_006238471.1 | 3 | 606 | ||
| XM_008763911.1 | XP_008762133.1 | 3 | 225 | |||
| XM_008763912.1 | XP_008762134.1 | 3 | 225 | |||
| XM_008763913.1 | XP_008762135.1 | 3 | 225 | |||
| XM_008763914.1 | XP_008762136.1 | 3 | 225 | |||

Mysm1
Genomic Map