siRNA Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106669.1 | NP_001100139.1 | 2 | 356 | ||
| XM_006238475.2 | XP_006238537.1 | 2 | 379 | |||
| XM_006238476.2 | XP_006238538.1 | 2 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oma1| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106669.1 | NP_001100139.1 | 2 | 356 | ||
| XM_006238475.2 | XP_006238537.1 | 2 | 379 | |||
| XM_006238476.2 | XP_006238538.1 | 2 | 370 | |||