siRNA Details
| Gene Symbol: C8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106670.1 | NP_001100140.1 | 6 | 707 | ||
| GenBank mRNA | FQ211202.1 | - | 6 | 786 | ||
| FQ211244.1 | - | 6 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8a| Gene Symbol: C8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106670.1 | NP_001100140.1 | 6 | 707 | ||
| GenBank mRNA | FQ211202.1 | - | 6 | 786 | ||
| FQ211244.1 | - | 6 | 786 | |||