siRNA Details
| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106671.1 | NP_001100141.1 | 7 | 1390 | ||
| XM_006238502.2 | XP_006238564.1 | 4 | 1942 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc22| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106671.1 | NP_001100141.1 | 7 | 1390 | ||
| XM_006238502.2 | XP_006238564.1 | 4 | 1942 | |||