siRNA Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139465.2 | NP_001132937.1 | 7 | 773 | ||
| GenBank mRNA | BF287725.1 | - | 3 | 122 | ||
| FM091711.1 | - | 3 | 193 | |||
| FM094601.1 | - | 6 | 505 | |||
| FQ212504.1 | - | 7 | 739 | |||
| FQ226828.1 | - | 6 | 493 | |||
| FQ232106.1 | - | 7 | 778 | |||

Tmem59
Genomic Map