siRNA Details
| Gene Symbol: ZNF79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286696.1 | NP_001273625.1 | 3 | 651 | ||
| NM_001286697.1 | NP_001273626.1 | 4 | 653 | |||
| NM_001322260.1 | NP_001309189.1 | 4 | 782 | |||
| NM_007135.2 | NP_009066.2 | 3 | 544 | |||
| XM_006717279.3 | XP_006717342.1 | 4 | 752 | |||
| GenBank mRNA | AK054606.1 | - | 3 | 536 | ||
| AK302284.1 | - | 3 | 593 | |||
| AK316233.1 | - | 4 | 589 | |||
| BC062309.1 | - | 3 | 499 | 3944657 | ||
| BP351823.1 | - | 3 | 506 | |||

ZNF79
Genomic Map