siRNA Details
| Gene Symbol: Rhox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024873.1 | NP_001020044.1 | 1 | 353 | ||
| GenBank mRNA | DQ058658.1 | - | 1 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox11| Gene Symbol: Rhox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024873.1 | NP_001020044.1 | 1 | 353 | ||
| GenBank mRNA | DQ058658.1 | - | 1 | 353 | ||