siRNA Details
| Gene Symbol: Rhox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024874.1 | NP_001020045.1 | 2 | 120 | ||
| GenBank mRNA | DQ058656.1 | - | 2 | 120 | ||
| Gene Symbol: LOC100364002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257483.2 | XP_006257545.1 | 2 | 112 | ||

Rhox9
Genomic Map