siRNA Details
| Gene Symbol: Rhox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025654.1 | NP_001020825.1 | 2 | 244 | ||
| GenBank mRNA | DQ058654.1 | - | 2 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox7| Gene Symbol: Rhox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025654.1 | NP_001020825.1 | 2 | 244 | ||
| GenBank mRNA | DQ058654.1 | - | 2 | 244 | ||