siRNA Details
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134519.1 | NP_001127991.1 | 1537 | |||
XM_006238584.2 | XP_006238646.1 | 1974 | ||||
XM_006238585.2 | XP_006238647.1 | 1564 | ||||
GenBank mRNA | BC168701.1 | - | 1553 | 7454685 |
Gene Symbol: LOC100910316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749994.3 | XP_003750042.1 | 16, 17 | 2639 | ||
XM_006238582.2 | XP_006238644.1 | 16, 17 | 1974 | |||
XM_008763953.1 | XP_008762175.1 | 16, 17 | 2280 |
