siRNA Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100840.1 | NP_001094310.1 | 4 | 407 | ||
| GenBank mRNA | BC100255.1 | - | 4 | 349 | ||
| BC158737.1 | - | 4 | 353 | |||
| CV117277.1 | - | 4 | 644 | |||
| FQ218354.1 | - | 4 | 304 | |||

Txndc12
Genomic Map