siRNA Details
| Gene Symbol: Coa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106674.1 | NP_001100144.1 | 2 | 272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coa7| Gene Symbol: Coa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106674.1 | NP_001100144.1 | 2 | 272 | ||