siRNA Details
| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013091.2 | NP_001013109.2 | 4 | 745 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gba2| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013091.2 | NP_001013109.2 | 4 | 745 | ||