siRNA Details
| Gene Symbol: Creb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013092.2 | NP_001013110.1 | 1, 2 | 338 | ||
| XM_006238050.1 | XP_006238112.1 | 1, 2 | 364 | |||
| XM_008763672.1 | XP_008761894.1 | 2, 3 | 168 | |||
| GenBank mRNA | BC062241.1 | - | 1, 2 | 341 | 6887761 | |
| FQ219084.1 | - | 1, 2 | 316 | |||
| KJ817427.1 | - | 1, 2 | 182 | |||

Creb3
Genomic Map