siRNA Details
| Gene Symbol: Arhgef39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106676.2 | NP_001100146.2 | 8 | 941 | ||
| GenBank mRNA | BC158673.1 | - | 8 | 941 | 8371589 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef39| Gene Symbol: Arhgef39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106676.2 | NP_001100146.2 | 8 | 941 | ||
| GenBank mRNA | BC158673.1 | - | 8 | 941 | 8371589 | |