siRNA Details
| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025655.1 | NP_001020826.1 | 2 | 346 | ||
| GenBank mRNA | BC098727.1 | - | 2 | 346 | 7379366 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025647, AK002526, AK150162, BC017684, AK004827, and others.

Cmpk1
Genomic Map