siRNA Details
| Gene Symbol: ZNF85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 813 | ||
| NM_001256173.1 | NP_001243102.1 | 3 | 596 | |||
| NM_003429.4 | NP_003420.2 | 4 | 723 | |||
| XM_011528263.2 | XP_011526565.1 | 6 | 884 | |||
| XM_011528264.1 | XP_011526566.1 | 2 | 1044 | |||
| XM_017027234.1 | XP_016882723.1 | 2 | 523 | |||
| GenBank mRNA | AK308258.1 | - | 4 | 553 | ||
| BC047646.1 | - | 2 | 1026 | 4828290 | ||
| M61866.1 | - | 4 | 687 | |||
| M61868.1 | - | 2 | 443 | |||
| U35376.1 | - | 4 | 693 | |||

ZNF85
Genomic Map