siRNA Details
| Gene Symbol: Efcab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106677.2 | NP_001100147.1 | 11 | 1687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab14| Gene Symbol: Efcab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106677.2 | NP_001100147.1 | 11 | 1687 | ||