siRNA Details
| Gene Symbol: Nsun4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106678.1 | NP_001100148.1 | 4 | 764 | ||
| Gene Symbol: LOC103690007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763980.1 | XP_008762202.1 | 1658 | |||

Nsun4
Genomic Map