siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025656.1 | NP_001020827.1 | 3 | 651 | ||
| XM_006238648.2 | XP_006238710.1 | 3 | 271 | |||
| XM_006238649.2 | XP_006238711.1 | 3 | 225 | |||
| XM_006238650.2 | XP_006238712.1 | 3 | 222 | |||
| XM_008763995.1 | XP_008762217.1 | 3 | 274 | |||
| GenBank mRNA | BC082064.1 | - | 3 | 730 | ||
| BC098949.1 | - | 3 | 651 | 7131942 | ||

Ccdc163
Genomic Map