siRNA Details
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106680.1 | NP_001100150.1 | 6 | 660 | ||
| XM_006238651.1 | XP_006238713.1 | 6 | 655 | |||
| XM_006238652.2 | XP_006238714.1 | 5 | 985 | |||
| XM_006238653.2 | XP_006238715.1 | 5 | 907 | |||
| XM_006238654.2 | XP_006238716.1 | 5 | 1008 | |||

Toe1
Genomic Map