siRNA Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015007.1 | NP_001015007.1 | 1 | 212 | ||
| GenBank mRNA | BC092656.1 | - | 1 | 212 | 7320904 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dph2| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015007.1 | NP_001015007.1 | 1 | 212 | ||
| GenBank mRNA | BC092656.1 | - | 1 | 212 | 7320904 | |