siRNA Details
| Gene Symbol: Olr865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000410.1 | NP_001000410.1 | 1 | 481 | ||
| XM_008763996.1 | XP_008762218.1 | 1 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr865| Gene Symbol: Olr865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000410.1 | NP_001000410.1 | 1 | 481 | ||
| XM_008763996.1 | XP_008762218.1 | 1 | 481 | |||