siRNA Details
| Gene Symbol: Olr865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000410.1 | NP_001000410.1 | 1 | 902 | ||
| XM_008763996.1 | XP_008762218.1 | 1 | 902 | |||
| Gene Symbol: Olr866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000411.1 | NP_001000411.1 | 1 | 902 | ||

Olr865
Genomic Map