siRNA Details
| Gene Symbol: Olr866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000411.1 | NP_001000411.1 | 1 | 481 | ||
| Gene Symbol: Olfr1330-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238754.1 | XP_006238816.1 | 2 | 556 | ||

Olr866
Genomic Map