siRNA Details
| Gene Symbol: Olr869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000413.1 | NP_001000413.1 | 1 | 902 | ||
| Gene Symbol: Olr867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000412.1 | NP_001000412.1 | 1 | 902 | ||

Olr869
Genomic Map