siRNA Details
| Gene Symbol: ZNF99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080409.2 | NP_001073878.2 | 4 | 1796 | ||
| Gene Symbol: ZNF729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242680.1 | NP_001229609.1 | 4 | 1906 | ||
| XM_017026122.1 | XP_016881611.1 | 4 | 1952 | |||

ZNF99
Genomic Map