siRNA Details
| Gene Symbol: Rragc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048184.1 | NP_001041649.1 | 1 | 236 | ||
| GenBank mRNA | BC105622.1 | - | 1 | 236 | 7454011 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rragc| Gene Symbol: Rragc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048184.1 | NP_001041649.1 | 1 | 236 | ||
| GenBank mRNA | BC105622.1 | - | 1 | 236 | 7454011 | |