siRNA Details
| Gene Symbol: Csf3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106685.1 | NP_001100155.1 | 6 | 752 | ||
| XM_008764003.1 | XP_008762225.1 | 6 | 990 | |||
| XM_008764004.1 | XP_008762226.1 | 5 | 899 | |||
| XM_008764005.1 | XP_008762227.1 | 6 | 818 | |||
| XM_008764006.1 | XP_008762228.1 | 5 | 727 | |||
| XM_008764007.1 | XP_008762229.1 | 6 | 990 | |||

Csf3r
Genomic Map