siRNA Details
| Gene Symbol: Dnali1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031647.1 | NP_001026817.1 | 5 | 727 | ||
| GenBank mRNA | BC099087.1 | - | 5 | 750 | 7455645 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnali1| Gene Symbol: Dnali1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031647.1 | NP_001026817.1 | 5 | 727 | ||
| GenBank mRNA | BC099087.1 | - | 5 | 750 | 7455645 | |