siRNA Details
| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106687.1 | NP_001100157.1 | 8 | 1272 | ||
| XM_006238863.2 | XP_006238925.1 | 8 | 1299 | |||
| XM_006238864.2 | XP_006238926.1 | 8 | 1299 | |||
| XM_008764008.1 | XP_008762230.1 | 8 | 1299 | |||
| Gene Symbol: LOC100912530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764129.1 | XP_008762351.1 | 6 | 1084 | ||

Clspn
Genomic Map