siRNA Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039011.1 | NP_001034100.1 | 17, 18 | 1553 | ||
| GenBank mRNA | BC099788.1 | - | 17, 18 | 1553 | 7934518 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnksr1| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039011.1 | NP_001034100.1 | 17, 18 | 1553 | ||
| GenBank mRNA | BC099788.1 | - | 17, 18 | 1553 | 7934518 | |