siRNA Details
| Gene Symbol: Tmem50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127525.1 | NP_001120997.1 | 3, 4 | 337 | ||
| GenBank mRNA | BC161895.1 | - | 3, 4 | 337 | 7304509 | |
| FQ213771.1 | - | 3, 4 | 325 | |||
| FQ220741.1 | - | 3, 4 | 321 | |||
| FQ225130.1 | - | 3, 4 | 329 | |||
| FQ235131.1 | - | 3, 4 | 321 | |||

Tmem50a
Genomic Map