siRNA Details
| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106690.1 | NP_001100160.1 | 8 | 1086 | ||
| XM_006239149.2 | XP_006239211.1 | 9 | 1345 | |||
| XM_008764216.1 | XP_008762438.1 | 9 | 1345 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grhl3| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106690.1 | NP_001100160.1 | 8 | 1086 | ||
| XM_006239149.2 | XP_006239211.1 | 9 | 1345 | |||
| XM_008764216.1 | XP_008762438.1 | 9 | 1345 | |||