siRNA Details
| Gene Symbol: ZNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008727.3 | NP_001008727.1 | 4 | 707 | ||
| NM_001308269.1 | NP_001295198.1 | 5 | 765 | |||
| XM_017027239.1 | XP_016882728.1 | 5 | 881 | |||
| GenBank mRNA | AK297919.1 | - | 5 | 764 | ||
| CN360954.1 | - | 3 | 653 | |||
| CN360955.1 | - | 1 | 122 | |||
| DC424100.1 | - | 5 | 763 | |||

ZNF121
Genomic Map